siRNA Details
Gene Symbol: COL2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001844.4 | NP_001835.3 | 54 | 4724 | ||
NM_033150.2 | NP_149162.2 | 53 | 4517 | |||
XM_017018828.1 | XP_016874317.1 | 56 | 4809 | |||
XM_017018829.1 | XP_016874318.1 | 56 | 4806 | |||
XM_017018830.1 | XP_016874319.1 | 55 | 4599 | |||
XM_017018831.1 | XP_016874320.1 | 54 | 6109 | |||
GenBank mRNA | AL834148.1 | - | 37 | 3629 | ||
BC007252.1 | - | 7 | 1219 | |||
BC116449.1 | - | 54 | 4545 | 7486698 | ||
X06268.1 | - | 5 | 1021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
