Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305134.1 | NP_001292063.1 | 4 | 835 | ||
NM_001305145.1 | NP_001292074.1 | 4 | 835 | |||
NM_016192.3 | NP_057276.2 | 4 | 835 | |||
XM_017003740.1 | XP_016859229.1 | 4 | 842 | |||
GenBank mRNA | AB004064.1 | - | 504 | |||
AB017269.1 | - | 4 | 793 | |||
AF179274.2 | - | 4 | 820 | |||
AF242222.1 | - | 4 | 835 | |||
AK054646.1 | - | 4 | 1615 | |||
AK074507.1 | - | 4 | 449 | |||
AK074632.1 | - | 4 | 818 | |||
AK222691.1 | - | 4 | 840 | |||
AK315376.1 | - | 4 | 836 | |||
AL157430.1 | - | 4 | 818 | |||
AY358907.1 | - | 4 | 801 | |||
BC008973.1 | - | 4 | 776 | 4180709 | ||
CR457390.1 | - | 4 | 426 | |||
DQ133599.1 | - | 4 | 426 |