Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016204.2 | NP_057288.1 | 2 | 689 | ||
GenBank mRNA | AF188285.1 | - | 2 | 673 | ||
AK314956.1 | - | 2 | 689 | |||
BC069643.1 | - | 2 | 592 | 7262433 | ||
BC074921.2 | - | 2 | 541 | 30915569 |