Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BNIP3 SIRNA Assays ›
          Gene Symbol:
          BNIP3
          Gene Name:
          BCL2 interacting protein 3
          Gene Aliases:
          NIP3
          Chromosome Location:
          Chr.10: 131967683 - 131982013 on Build GRCh38
          UniGene ID:
          Hs.144873
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 241440

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BNIP3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004052.3 NP_004043.3 3, 4 482
          GenBank mRNA AF002697.1 - 3, 4 401
          AK222626.1 - 3, 4 392
          AK295136.1 - 3, 4 392
          BC009342.2 - 3, 4 377
          BC021989.1 - 3, 4 396 4066029
          BC067818.1 - 3, 4 425
          BC080643.1 - 3, 4 377
          U15174.1 - 3, 4 346

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy
          response to hypoxia
          apoptotic process
          cell death
          granzyme-mediated apoptotic signaling pathway
          positive regulation of autophagy
          negative regulation of mitochondrial fusion
          positive regulation of cardiac muscle cell apoptotic process
          positive regulation of macroautophagy
          mitochondrial protein catabolic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of programmed cell death
          positive regulation of protein complex disassembly
          mitochondrial fragmentation involved in apoptotic process
          negative regulation of membrane potential
          regulation of mitochondrial membrane permeability
          autophagic cell death
          brown fat cell differentiation
          neuron apoptotic process
          defense response to virus
          response to hyperoxia
          negative regulation of cell death
          cellular response to hydrogen peroxide
          cellular response to mechanical stimulus
          cellular response to cobalt ion
          cellular response to hypoxia
          reactive oxygen species metabolic process
          positive regulation of mitochondrial fission
          positive regulation of release of cytochrome c from mitochondria
          mitochondrial outer membrane permeabilization
          toxin transport
          regulation of aerobic respiration
          intrinsic apoptotic signaling pathway in response to hypoxia
          negative regulation of reactive oxygen species metabolic process
          protein binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0