Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYH10 SIRNA Assays ›
          Gene Symbol:
          MYH10
          Gene Name:
          myosin, heavy chain 10, non-muscle
          Gene Aliases:
          NMMHC-IIB, NMMHCB
          Chromosome Location:
          Chr.17: 8474205 - 8631369 on Build GRCh38
          UniGene ID:
          Hs.16355
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 241476

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYH10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256012.1 NP_001242941.1 8, 9 957
          NM_001256095.1 NP_001243024.1 8, 9 954
          NM_005964.3 NP_005955.3 7, 8 927
          XM_011523875.2 XP_011522177.1 8, 9 1246
          XM_011523878.2 XP_011522180.1 8, 9 1298
          XM_011523879.2 XP_011522181.1 8, 9 1243
          XM_011523880.2 XP_011522182.1 7, 8 1216
          XM_017024677.1 XP_016880166.1 8, 9 1012
          XM_017024678.1 XP_016880167.1 9, 10 994
          XM_017024679.1 XP_016880168.1 9, 10 1035
          XM_017024680.1 XP_016880169.1 9, 10 1343
          XM_017024681.1 XP_016880170.1 8, 9 964
          XM_017024682.1 XP_016880171.1 8, 9 939
          GenBank mRNA AB210026.1 - 8, 9 909
          BC117690.1 - 6, 7 829 40072059
          BC117691.1 - 6, 7 829 40072061
          BC144668.1 - 8, 9 911 9053199
          BC150634.1 - 8, 9 911 9057004
          BG421687.1 - 3, 4 198
          M69181.1 - 7, 8 867
          U15618.1 - 4, 5 203

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031520, XM_008767880, XM_008767881, XM_006246821, AF139055, and others.
          This siRNA also targets the following transcripts in Mouse: NM_175260, AK045066, AK147567, AK045074, AK133946, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          in utero embryonic development
          neuron migration
          plasma membrane repair
          cardiac septum development
          exocytosis
          substrate-dependent cell migration, cell extension
          nuclear migration
          cell adhesion
          axon guidance
          adult heart development
          cell proliferation
          regulation of cell shape
          fourth ventricle development
          lateral ventricle development
          third ventricle development
          cerebellar Purkinje cell layer development
          actin filament-based movement
          actomyosin structure organization
          aorta development
          neuromuscular process controlling balance
          cardiac myofibril assembly
          ventricular cardiac muscle cell development
          retina development in camera-type eye
          coronary vasculature development
          microfilament motor activity
          actin binding
          protein binding
          calmodulin binding
          ATP binding
          actin-dependent ATPase activity
          ADP binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0