Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318046.1 | NP_001304975.1 | 23 | 3338 | ||
NM_001318047.1 | NP_001304976.1 | 23 | 3341 | |||
NM_007356.2 | NP_031382.2 | 23 | 3341 | |||
XM_011515975.1 | XP_011514277.1 | 23 | 3341 | |||
XM_011515978.1 | XP_011514280.1 | 23 | 3341 | |||
XM_011515979.2 | XP_011514281.1 | 23 | 3341 | |||
XM_011515980.2 | XP_011514282.1 | 23 | 3341 | |||
XM_017011879.1 | XP_016867368.1 | 23 | 3341 | |||
XM_017011880.1 | XP_016867369.1 | 23 | 3341 | |||
GenBank mRNA | AF028816.1 | - | 23 | 3337 | ||
AF029325.1 | - | 1 | 109 | |||
AK300385.1 | - | 23 | 3342 | |||
AK301777.1 | - | 2 | 333 | |||
BC045172.2 | - | 2 | 335 | |||
BC140804.1 | - | 23 | 3328 | 9021672 | ||
BC144591.1 | - | 23 | 3328 | 9053121 |