Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCC1 SIRNA Assays ›
Gene Symbol:
TMCC1
Gene Name:
transmembrane and coiled-coil domain family 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 129647792 - 129893605 on Build GRCh38
UniGene ID:
Hs.477547
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241574
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001017395.3
NP_001017395.2
4
1866
NM_001128224.2
NP_001121696.1
4
1267
NR_033361.1
2
1412
XM_006713542.3
XP_006713605.1
5
1979
XM_006713543.3
XP_006713606.1
5
1976
XM_006713544.1
XP_006713607.1
5
2104
XM_006713550.3
XP_006713613.1
5
1537
XM_006713552.3
XP_006713615.1
2
1159
XM_006713553.3
XP_006713616.1
2
2451
XM_011512572.2
XP_011510874.1
6
2139
XM_011512573.2
XP_011510875.1
5
2718
XM_011512576.2
XP_011510878.1
5
1972
XM_011512579.2
XP_011510881.1
4
1727
XM_011512580.2
XP_011510882.1
7
1864
XM_011512581.2
XP_011510883.1
6
1701
XM_011512582.1
XP_011510884.1
2
1493
XM_011512583.2
XP_011510885.1
4
1270
XM_011512584.2
XP_011510886.1
6
1589
XM_017005931.1
XP_016861420.1
5
1887
XM_017005932.1
XP_016861421.1
6
2126
XM_017005933.1
XP_016861422.1
4
1923
XM_017005934.1
XP_016861423.1
8
1937
XM_017005935.1
XP_016861424.1
9
2026
XM_017005936.1
XP_016861425.1
6
1700
XM_017005937.1
XP_016861426.1
6
1634
XM_017005938.1
XP_016861427.1
8
2000
XM_017005939.1
XP_016861428.1
7
1894
XM_017005940.1
XP_016861429.1
4
1272
XM_017005941.1
XP_016861430.1
4
1398
GenBank mRNA
AB018322.1
-
1
433
AK124671.1
-
2
1413
AK291448.1
-
4
1685
AK293408.1
-
2
949
BC030619.2
-
2
1330
BC034284.1
-
1
16
BC039859.1
-
4
1773
5527623
BC047517.1
-
2
1330
BX640712.1
-
2
920
CR749206.1
-
2
1392
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0