Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260491.1 | NP_001247420.1 | 6 | 726 | ||
NM_015266.2 | NP_056081.1 | 6 | 726 | |||
NR_048537.1 | 6 | 726 | ||||
NR_048538.1 | 5 | 642 | ||||
NR_048539.1 | 6 | 726 | ||||
XM_006723756.1 | XP_006723819.1 | 6 | 588 | |||
XM_011528736.1 | XP_011527038.1 | 6 | 588 | |||
XM_011528737.1 | XP_011527039.1 | 5 | 461 | |||
XM_011528738.1 | XP_011527040.1 | 6 | 588 | |||
XM_011528739.1 | XP_011527041.1 | 6 | 588 | |||
XM_011528740.1 | XP_011527042.1 | 6 | 588 | |||
XM_011528741.1 | XP_011527043.1 | 6 | 588 | |||
XM_011528742.1 | XP_011527044.1 | 5 | 444 | |||
XM_011528745.1 | XP_011527047.1 | 6 | 588 | |||
XM_017027754.1 | XP_016883243.1 | 6 | 588 | |||
XM_017027755.1 | XP_016883244.1 | 6 | 532 | |||
XM_017027756.1 | XP_016883245.1 | 6 | 532 | |||
GenBank mRNA | AB023156.2 | - | 6 | 558 | ||
AK295665.1 | - | 5 | 444 | |||
AK295803.1 | - | 7 | 529 | |||
AK295822.1 | - | 6 | 519 | |||
AK300318.1 | - | 6 | 611 | |||
AK310530.1 | - | 6 | 528 | |||
BC112213.1 | - | 6 | 548 | 8327681 | ||
BX430866.2 | - | 1 | 52 | |||
CR749638.1 | - | 6 | 603 |