Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260491.1 | NP_001247420.1 | 16 | 4407 | ||
NM_015266.2 | NP_056081.1 | 16 | 4359 | |||
NR_048537.1 | 15 | 4324 | ||||
NR_048538.1 | 14 | 4240 | ||||
NR_048539.1 | 14 | 4196 | ||||
NR_048540.1 | 12 | 4022 | ||||
XM_006723756.1 | XP_006723819.1 | 17 | 4320 | |||
XM_011528736.1 | XP_011527038.1 | 17 | 4368 | |||
XM_011528737.1 | XP_011527039.1 | 16 | 4241 | |||
XM_011528738.1 | XP_011527040.1 | 16 | 4251 | |||
XM_011528739.1 | XP_011527041.1 | 16 | 4245 | |||
XM_011528741.1 | XP_011527043.1 | 15 | 4152 | |||
XM_011528742.1 | XP_011527044.1 | 15 | 4141 | |||
XM_011528743.2 | XP_011527045.1 | 9 | 3505 | |||
XM_011528744.2 | XP_011527046.1 | 8 | 3336 | |||
XM_017027755.1 | XP_016883244.1 | 16 | 4229 | |||
XM_017027756.1 | XP_016883245.1 | 17 | 4317 | |||
GenBank mRNA | AB023156.2 | - | 16 | 4191 | ||
AL049469.1 | - | 1 | 428 | |||
AL833574.1 | - | 5 | 3058 |