Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017934.5 | NP_060404.3 | 35 | 4176 | ||
XM_005248729.4 | XP_005248786.1 | 35 | 4173 | |||
XM_011535918.2 | XP_011534220.1 | 32 | 3832 | |||
XM_017010989.1 | XP_016866478.1 | 37 | 4318 | |||
XM_017010990.1 | XP_016866479.1 | 34 | 3916 | |||
GenBank mRNA | AF310250.1 | - | 12 | 1183 | ||
AJ303102.1 | - | 35 | 3963 | |||
AK000712.1 | - | 8 | 806 | |||
AK075124.1 | - | 4 | 263 | |||
AL161957.1 | - | 12 | 4030 | |||
BC008909.2 | - | 10 | 904 | |||
BC064611.1 | - | 7 | 645 | |||
BC081569.1 | - | 13 | 1219 | |||
BC137488.1 | - | 35 | 4056 | 9021488 | ||
DQ924532.1 | - | 35 | 3950 |