Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R3 SIRNA Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location:
Chr.11: 68460712 - 68615334 on Build GRCh38
UniGene ID:
Hs.503022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241719
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164160.1
NP_001157632.1
20
2320
NM_001164161.1
NP_001157633.1
20
2320
NM_001164162.1
NP_001157634.1
20
2302
NM_001164163.1
NP_001157635.1
19
2233
NM_001164164.1
NP_001157636.1
19
2185
NM_018312.4
NP_060782.2
19
2185
XM_006718608.3
XP_006718671.1
21
2436
XM_006718612.3
XP_006718675.1
21
2418
XM_006718613.2
XP_006718676.1
21
2436
XM_006718614.2
XP_006718677.1
21
2418
XM_006718618.3
XP_006718681.1
20
2349
XM_006718621.2
XP_006718684.1
20
2298
XM_006718622.3
XP_006718685.1
19
2196
XM_006718623.3
XP_006718686.1
19
2179
XM_006718624.2
XP_006718687.1
19
2196
XM_011545135.1
XP_011543437.1
21
2436
XM_011545136.2
XP_011543438.1
20
2319
XM_011545137.2
XP_011543439.1
21
2419
XM_011545138.2
XP_011543440.1
22
2637
XM_011545139.1
XP_011543441.1
21
2418
XM_011545140.2
XP_011543442.1
21
2436
XM_011545141.1
XP_011543443.1
21
2385
XM_011545142.2
XP_011543444.1
20
2349
XM_011545143.1
XP_011543445.1
20
2331
XM_011545145.2
XP_011543447.1
20
2283
XM_017017962.1
XP_016873451.1
20
2274
XM_017017963.1
XP_016873452.1
21
2360
XM_017017964.1
XP_016873453.1
21
2418
XM_017017965.1
XP_016873454.1
20
2301
XM_017017966.1
XP_016873455.1
21
2504
XM_017017967.1
XP_016873456.1
21
2385
XM_017017968.1
XP_016873457.1
20
2268
XM_017017969.1
XP_016873458.1
21
2385
XM_017017970.1
XP_016873459.1
20
2268
XM_017017971.1
XP_016873460.1
20
2349
XM_017017972.1
XP_016873461.1
19
2205
XM_017017973.1
XP_016873462.1
20
2332
XM_017017974.1
XP_016873463.1
20
2331
XM_017017975.1
XP_016873464.1
20
2298
XM_017017976.1
XP_016873465.1
19
2115
XM_017017977.1
XP_016873466.1
20
2207
XM_017017978.1
XP_016873467.1
19
2058
XM_017017979.1
XP_016873468.1
19
2040
XM_017017980.1
XP_016873469.1
19
2058
XM_017017981.1
XP_016873470.1
19
2007
XM_017017982.1
XP_016873471.1
18
1971
XM_017017983.1
XP_016873472.1
20
2457
XM_017017984.1
XP_016873473.1
19
2300
GenBank mRNA
AB046778.2
-
20
2268
AF264779.1
-
18
2090
AF264780.1
-
18
2090
AK023950.1
-
3
226
AK057250.1
-
14
1393
AK125055.1
-
5
2072
AK300359.1
-
12
1154
AK302611.1
-
12
1174
AK303315.1
-
15
1636
AK315136.1
-
19
2202
AL834471.1
-
3
197
BC007738.2
-
2
108
BC020953.1
-
6
442
BC105933.1
-
19
2188
40029064
BC105934.1
-
19
2188
40029058
BC107599.1
-
18
2101
40029065
CR749815.1
-
20
2265
CR933658.1
-
20
2283
CR936718.1
-
20
2288
DQ111954.2
-
18
2053
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of phosphoprotein phosphatase activity
COPII vesicle coating
protein binding
protein phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0