Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032048.2 | NP_114437.2 | 8 | 3475 | ||
GenBank mRNA | AB026706.1 | - | 4 | 781 | ||
AF270513.1 | - | 8 | 3475 | |||
AK090519.1 | - | 5 | 1570 | |||
BC032231.1 | - | 6 | 1148 | |||
BC136541.1 | - | 8 | 3429 | 9020531 |