Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BMPR1A SIRNA Assays ›
          Gene Symbol:
          BMPR1A
          Gene Name:
          bone morphogenetic protein receptor type 1A
          Gene Aliases:
          10q23del, ACVRLK3, ALK3, CD292, SKR5
          Chromosome Location:
          Chr.10: 86755786 - 86927969 on Build GRCh38
          UniGene ID:
          Hs.524477
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 242306

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BMPR1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004329.2 NP_004320.2 13 3339
          XM_011540103.2 XP_011538405.1 13 3236
          XM_011540104.2 XP_011538406.1 14 3403
          GenBank mRNA BC028383.1 - 13 3333 4824378
          DQ786215.1 - 1 398

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          mesoderm formation
          somitogenesis
          Mullerian duct regression
          positive regulation of mesenchymal cell proliferation
          chondrocyte differentiation
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          cardiac conduction system development
          mitral valve morphogenesis
          tricuspid valve morphogenesis
          endocardial cushion morphogenesis
          cardiac right ventricle morphogenesis
          ventricular trabecula myocardium morphogenesis
          ventricular compact myocardium morphogenesis
          endocardial cushion formation
          protein phosphorylation
          immune response
          transforming growth factor beta receptor signaling pathway
          ectoderm development
          dorsal/ventral axis specification
          regulation of cardiac muscle cell apoptotic process
          positive regulation of pathway-restricted SMAD protein phosphorylation
          neural crest cell development
          negative regulation of smooth muscle cell migration
          stem cell population maintenance
          pituitary gland development
          neural plate mediolateral regionalization
          signal transduction by protein phosphorylation
          lung development
          positive regulation of bone mineralization
          BMP signaling pathway
          hindlimb morphogenesis
          dorsal aorta morphogenesis
          odontogenesis of dentin-containing tooth
          embryonic digit morphogenesis
          positive regulation of osteoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          paraxial mesoderm structural organization
          lateral mesoderm development
          regulation of lateral mesodermal cell fate specification
          mesendoderm development
          embryonic organ development
          developmental growth
          positive regulation of epithelial cell proliferation
          negative regulation of neurogenesis
          palate development
          regulation of cardiac muscle cell proliferation
          positive regulation of cardiac muscle cell proliferation
          positive regulation of SMAD protein import into nucleus
          heart formation
          pharyngeal arch artery morphogenesis
          cellular response to BMP stimulus
          positive regulation of cardiac ventricle development
          positive regulation of vascular smooth muscle cell proliferation
          regulation of cellular senescence
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          glycoprotein binding
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          BMP receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0