Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042452.1 | NP_001035917.1 | 5 | 484 | ||
NM_001042453.1 | NP_001035918.1 | 5 | 544 | |||
NM_016542.3 | NP_057626.2 | 6 | 775 | |||
XM_011531349.1 | XP_011529651.1 | 7 | 748 | |||
XM_011531350.1 | XP_011529652.1 | 6 | 821 | |||
GenBank mRNA | AB040057.2 | - | 6 | 694 | ||
AF231012.1 | - | 6 | 663 | |||
AF344882.1 | - | 5 | 484 | |||
AF344883.1 | - | 5 | 484 | |||
AK000556.1 | - | 2 | 107 | |||
AK074837.1 | - | 1 | 28 | |||
AK075107.1 | - | 5 | 456 | |||
AK075150.1 | - | 6 | 690 | |||
AK303583.1 | - | 7 | 756 | |||
AK314356.1 | - | 6 | 691 | |||
AK316137.1 | - | 7 | 756 | |||
BC098315.1 | - | 6 | 680 | 40026349 | ||
BC099843.1 | - | 6 | 680 | 40026348 | ||
BC103503.1 | - | 6 | 680 | 40026350 | ||
BT020099.1 | - | 5 | 474 |