Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003565.2 | NP_003556.1 | 28 | 4796 | ||
XM_011538798.2 | XP_011537100.1 | 28 | 4642 | |||
XM_011538799.2 | XP_011537101.1 | 28 | 4555 | |||
GenBank mRNA | AB018265.2 | - | 28 | 4578 | ||
AF045458.1 | - | 28 | 4796 | |||
AK095774.1 | - | 2 | 1921 | |||
AK128333.1 | - | 1 | 2623 | |||
BI822269.1 | - | 1 | 200 |