Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AURKB SIRNA Assays ›
          Gene Symbol:
          AURKB
          Gene Name:
          aurora kinase B
          Gene Aliases:
          AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2
          Chromosome Location:
          Chr.17: 8204731 - 8210768 on Build GRCh38
          UniGene ID:
          Hs.442658
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 242382

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AURKB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256834.2 NP_001243763.1 4 311
          NM_001284526.1 NP_001271455.1 5 387
          NM_001313950.1 NP_001300879.1 5 360
          NM_001313951.1 NP_001300880.1 5 389
          NM_001313952.1 NP_001300881.1 4 375
          NM_001313954.1 NP_001300883.1 5 384
          NM_004217.3 NP_004208.2 5 384
          NR_132731.1 4 266
          XM_011524072.2 XP_011522374.1 4 500
          XM_017025307.1 XP_016880796.1 4 427
          XM_017025309.1 XP_016880798.1 4 255
          GenBank mRNA AB011446.1 - 5 328
          AB011450.1 - 5 319
          AB519677.1 - 4 262
          AB519679.1 - 4 265
          AF004022.1 - 5 320
          AF008552.1 - 4 262
          AF015254.1 - 5 320
          AK297976.1 - 4 266
          AK314889.1 - 5 333
          AY677083.1 - 4 266
          BC000442.2 - 5 323 2820719
          BC009751.2 - 5 303 3677349
          BC013300.2 - 5 343
          BC080581.1 - 5 315 2819846
          BE514591.1 - 4 309
          BM905813.1 - 4 336
          BP225027.1 - 5 370
          BT019534.1 - 4 262
          BU181480.1 - 4 243
          CX781462.1 - 5 296

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          negative regulation of B cell apoptotic process
          protein phosphorylation
          spindle organization
          sister chromatid cohesion
          aging
          cell proliferation
          attachment of spindle microtubules to kinetochore
          abscission
          histone modification
          protein sumoylation
          anaphase-promoting complex-dependent catabolic process
          spindle checkpoint
          negative regulation of protein binding
          positive regulation of telomere maintenance via telomerase
          negative regulation of cytokinesis
          positive regulation of cytokinesis
          protein localization to kinetochore
          cellular response to UV
          cleavage furrow formation
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          histone H3-S28 phosphorylation
          protein autophosphorylation
          mitotic spindle midzone assembly
          positive regulation of telomerase activity
          regulation of chromosome segregation
          regulation of signal transduction by p53 class mediator
          positive regulation of telomere capping
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          histone serine kinase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0