siRNA Details
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014.4 | NP_001005.1 | 6 | 627 | ||
NM_001203245.2 | NP_001190174.1 | 6 | 779 | |||
NM_001204091.1 | NP_001191020.1 | 6 | 631 | |||
GenBank mRNA | BC001032.2 | - | 6 | 552 | 3139393 | |
BC001955.1 | - | 6 | 606 | 2905318 | ||
BC005012.1 | - | 6 | 606 | 3637658 | ||
BC071946.1 | - | 6 | 562 | 6305105 | ||
BC073799.1 | - | 6 | 639 | 5517455 | ||
BE397113.1 | - | 6 | 577 | |||
BU956627.1 | - | 6 | 555 | |||
CD385233.1 | - | 6 | 629 | |||
U14972.1 | - | 1 | 549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
