siRNA Details
Gene Symbol: SOX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006942.1 | NP_008873.1 | 2 | 1084 | ||
GenBank mRNA | AB006867.1 | - | 2 | 1084 | ||
AJ006222.1 | - | 2 | 319 | |||
BC000985.2 | - | 2 | 988 | 3447385 | ||
BC072003.1 | - | 3 | 738 | 4653140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
