siRNA Details
Gene Symbol: NFIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271043.2 | NP_001257972.1 | 3 | 676 | ||
NM_001271044.2 | NP_001257973.1 | 3 | 636 | |||
NM_002501.3 | NP_002492.2 | 3 | 667 | |||
XM_005259917.4 | XP_005259974.1 | 3 | 954 | |||
XM_005259918.4 | XP_005259975.1 | 3 | 674 | |||
XM_005259919.3 | XP_005259976.1 | 3 | 791 | |||
XM_005259920.3 | XP_005259977.1 | 3 | 1353 | |||
XM_006722760.2 | XP_006722823.1 | 3 | 791 | |||
XM_011528040.1 | XP_011526342.1 | 3 | 674 | |||
XM_017026837.1 | XP_016882326.1 | 3 | 925 | |||
GenBank mRNA | AK295290.1 | - | 3 | 676 | ||
AK297261.1 | - | 635 | ||||
BC117113.1 | - | 2 | 630 | 40125664 | ||
BC117115.1 | - | 2 | 630 | 40125666 | ||
BT019732.1 | - | 3 | 599 | |||
BX537794.1 | - | 3 | 673 | |||
FJ861276.1 | - | 2 | 575 | |||
L31881.1 | - | 2 | 635 | |||
U07811.1 | - | 2 | 546 | |||
U18759.1 | - | 3 | 616 | |||
U18760.1 | - | 2 | 258 | |||
U18761.1 | - | 2 | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
