siRNA Details
Gene Symbol: FAM20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020223.3 | NP_064608.2 | 1 | 401 | ||
XM_017012450.1 | XP_016867939.1 | 1 | 396 | |||
XM_017012451.1 | XP_016867940.1 | 1 | 396 | |||
XM_017012452.1 | XP_016867941.1 | 1 | 396 | |||
XM_017012453.1 | XP_016867942.1 | 1 | 431 | |||
XM_017012454.1 | XP_016867943.1 | 1 | 411 | |||
GenBank mRNA | AB545605.1 | - | 1 | 401 | ||
AF533706.1 | - | 1 | 401 | |||
BC040074.1 | - | 1 | 257 | 4942737 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
