Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 3 | 283 | ||
NM_201400.3 | NP_958802.1 | 5 | 466 | |||
NM_201598.3 | NP_963892.1 | 4 | 364 | |||
XM_005255157.4 | XP_005255214.1 | 4 | 358 | |||
XM_005255158.4 | XP_005255215.1 | 6 | 512 | |||
XM_011522404.2 | XP_011520706.1 | 5 | 431 | |||
GenBank mRNA | AL522535.3 | - | 3 | 205 | ||
AY037162.1 | - | 4 | 332 | |||
BC010084.2 | - | 5 | 386 | 2822323 | ||
BG024981.1 | - | 4 | 332 | |||
BQ921410.1 | - | 5 | 449 | |||
CA495371.1 | - | 4 | 285 | |||
HY017433.1 | - | 4 | 364 | |||
JX424572.1 | - | 2 | 74 |