Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001933.1 | NP_001001933.1 | 6 | 1136 | ||
NM_001256114.1 | NP_001243043.1 | 5 | 527 | |||
XM_017001316.1 | XP_016856805.1 | 5 | 1016 | |||
XM_017001317.1 | XP_016856806.1 | 4 | 536 | |||
GenBank mRNA | BC040321.1 | - | 6 | 1136 | 4839343 |