siRNA Details
Gene Symbol: POLR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137604.2 | NP_001131076.1 | 14 | 3432 | ||
NM_001282772.1 | NP_001269701.1 | 16 | 3250 | |||
NM_001282774.1 | NP_001269703.1 | 13 | 3051 | |||
NM_001282776.1 | NP_001269705.1 | 12 | 3152 | |||
NM_001282777.1 | NP_001269706.1 | 16 | 3692 | |||
NM_001282779.1 | NP_001269708.1 | 15 | 3529 | |||
NM_019014.5 | NP_061887.2 | 15 | 3600 | |||
GenBank mRNA | AK022533.1 | - | 14 | 2882 | ||
AK022890.1 | - | 8 | 1763 | |||
AK025574.1 | - | 4 | 1108 | |||
AK128044.1 | - | 15 | 3600 | |||
AK172817.1 | - | 1 | 437 | |||
AK294027.1 | - | 15 | 3001 | |||
AK301178.1 | - | 12 | 2624 | |||
AK302814.1 | - | 16 | 3250 | |||
BC004882.2 | - | 6 | 1362 | |||
BC073857.1 | - | 6 | 1296 | |||
BC096748.1 | - | 6 | 1397 | |||
BC110833.1 | - | 13 | 2740 | 6053210 | ||
BC150139.1 | - | 7 | 1806 | |||
BX647360.1 | - | 16 | 3418 | |||
BX647683.1 | - | 15 | 2979 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -





