Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094396.1 | - | 1 | 302 | ||
BC073926.1 | - | 1 | 383 |
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516187.2 | XP_011514489.1 | 1 | 32 | ||
XM_017012197.1 | XP_016867686.1 | 8 | 1331 |