siRNA Details
Gene Symbol: NLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014445.2 | NP_001014445.1 | 2, 3 | 487 | ||
NM_018096.4 | NP_060566.2 | 3, 4 | 416 | |||
GenBank mRNA | AB209111.1 | - | 2, 3 | 487 | ||
AK001320.1 | - | 3, 4 | 405 | |||
AK303253.1 | - | 3, 4 | 391 | |||
AK304508.1 | - | 3, 4 | 404 | |||
BC002884.2 | - | 2, 3 | 374 | |||
BC012075.1 | - | 3, 4 | 389 | 4652713 | ||
BI752930.1 | - | 2, 3 | 509 | |||
BI911995.1 | - | 3, 4 | 397 | |||
BP244661.1 | - | 3, 4 | 416 | |||
BQ070427.1 | - | 1, 2 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
