siRNA Details
Gene Symbol: APOA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000482.3 | NP_000473.2 | 3 | 465 | ||
GenBank mRNA | AL531739.3 | - | 3 | 465 | ||
BC074764.2 | - | 3 | 399 | 30915177 | ||
BC113594.1 | - | 3 | 390 | 8322646 | ||
BC113596.1 | - | 3 | 390 | 8322648 | ||
M13654.1 | - | 3 | 412 | |||
M14566.1 | - | 2 | 301 | |||
X13629.1 | - | 3 | 406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
