Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000482.3 | NP_000473.2 | 3 | 730 | ||
GenBank mRNA | AL531739.3 | - | 3 | 730 | ||
BC074764.2 | - | 3 | 664 | 30915177 | ||
BC113594.1 | - | 3 | 655 | 8322646 | ||
BC113596.1 | - | 3 | 655 | 8322648 | ||
M10373.1 | - | 1 | 224 | |||
M13654.1 | - | 3 | 677 | |||
M14566.1 | - | 2 | 566 | |||
X13629.1 | - | 3 | 671 |