siRNA Details
Gene Symbol: APOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000039.2 | NP_000030.1 | 3 | 421 | ||
NM_001318017.1 | NP_001304946.1 | 3 | 134 | |||
NM_001318018.1 | NP_001304947.1 | 3 | 126 | |||
GenBank mRNA | A14829.1 | - | 2 | 74 | ||
AK292231.1 | - | 3 | 95 | |||
AL531353.3 | - | 3 | 126 | |||
AL557908.3 | - | 3 | 134 | |||
BC005380.1 | - | 3 | 93 | 3934992 | ||
BC110286.1 | - | 3 | 116 | 5164525 | ||
CD558050.1 | - | 3 | 421 | |||
M11791.1 | - | 2 | 55 | |||
M27875.1 | - | 2 | 74 | |||
T68127.1 | - | 3 | 134 | |||
X00566.1 | - | 2 | 90 | |||
X02162.1 | - | 3 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
