siRNA Details
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014.4 | NP_001005.1 | 2 | 152 | ||
NM_001203245.2 | NP_001190174.1 | 2 | 304 | |||
NM_001204091.1 | NP_001191020.1 | 2 | 156 | |||
GenBank mRNA | AB209165.1 | - | 1 | 107 | ||
AK311797.1 | - | 2 | 99 | |||
BC001032.2 | - | 2 | 77 | 3139393 | ||
BC001955.1 | - | 2 | 131 | 2905318 | ||
BC005012.1 | - | 2 | 131 | 3637658 | ||
BC070235.1 | - | 2 | 75 | 6453788 | ||
BC071946.1 | - | 2 | 87 | 6305105 | ||
BC073799.1 | - | 2 | 164 | 5517455 | ||
BE397113.1 | - | 2 | 102 | |||
BG532491.1 | - | 2 | 305 | |||
BG940847.1 | - | 2 | 152 | |||
BQ072109.1 | - | 1 | 272 | |||
BU956627.1 | - | 2 | 80 | |||
BY996439.1 | - | 2 | 311 | |||
CD385233.1 | - | 2 | 154 | |||
U14972.1 | - | 1 | 74 |
Gene Symbol: RPS10-NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202470.2 | NP_001189399.1 | 2 | 152 | ||
GenBank mRNA | BU521288.1 | - | 2 | 75 |
Gene Symbol: RPS10P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025353.1 | - | 1 | 2686 |
Gene Symbol: RPS10P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026667.1 | 1 | 210 | |||
GenBank mRNA | AK055590.1 | - | 1 | 1408 | ||
BC030568.2 | - | 1 | 142 |
Gene Symbol: TFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU539534.1 | - | 772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
