siRNA Details
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014.4 | NP_001005.1 | 2 | 123 | ||
NM_001203245.2 | NP_001190174.1 | 2 | 275 | |||
NM_001204091.1 | NP_001191020.1 | 2 | 127 | |||
GenBank mRNA | AB209165.1 | - | 1 | 78 | ||
AK311797.1 | - | 2 | 70 | |||
BC001032.2 | - | 2 | 48 | 3139393 | ||
BC001955.1 | - | 2 | 102 | 2905318 | ||
BC005012.1 | - | 2 | 102 | 3637658 | ||
BC070235.1 | - | 2 | 46 | 6453788 | ||
BC071946.1 | - | 2 | 58 | 6305105 | ||
BC073799.1 | - | 2 | 135 | 5517455 | ||
BE397113.1 | - | 2 | 73 | |||
BG532491.1 | - | 2 | 276 | |||
BG940847.1 | - | 2 | 123 | |||
BQ072109.1 | - | 1 | 243 | |||
BU956627.1 | - | 2 | 51 | |||
BY996439.1 | - | 2 | 282 | |||
CD385233.1 | - | 2 | 125 |
Gene Symbol: RPS10-NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202470.2 | NP_001189399.1 | 2 | 123 | ||
GenBank mRNA | BU521288.1 | - | 2 | 46 |
Gene Symbol: RPS10P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025353.1 | - | 1 | 2657 |
Gene Symbol: TFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU539534.1 | - | 743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
