Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 7 | 949 | |||
GenBank mRNA | BC034017.1 | - | 7 | 950 | ||
BC067351.1 | - | 5 | 647 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027026.1 | 4 | 359 | |||
GenBank mRNA | AK095656.1 | - | 4 | 283 | ||
DA358891.1 | - | 4 | 359 |
Gene Symbol: GUSBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024448.2 | 11 | 2930 | |||
GenBank mRNA | AK056875.1 | - | 11 | 2860 | ||
AK091837.1 | - | 10 | 2668 | |||
AK125808.1 | - | 4 | 3330 | |||
BC063045.1 | - | 9 | 1173 | |||
BX538181.1 | - | 11 | 2823 | |||
CR933599.1 | - | 3 | 3202 | |||
S82637.1 | - | 3 | 422 | |||
X52204.1 | - | 3 | 424 |
Gene Symbol: LOC653080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Z70705.1 | - | 1 | 39 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 4 | 553 | |||
NR_029426.1 | 7 | 845 | ||||
GenBank mRNA | AK097091.1 | - | 8 | 1225 | ||
AK289832.1 | - | 7 | 927 | |||
AK315849.1 | - | 7 | 918 | |||
X75940.1 | - | 4 | 554 | |||
Z70701.1 | - | 1 | 39 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289851.1 | - | 7 | 847 |