siRNA Details
Gene Symbol: IFNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000605.3 | NP_000596.2 | 1 | 237 | ||
GenBank mRNA | AY255838.1 | - | 1 | 103 | ||
BC074936.2 | - | 1 | 213 | 30915269 | ||
BC074937.2 | - | 1 | 213 | 30915492 | ||
BC104163.1 | - | 1 | 214 | 40029595 | ||
BC104164.1 | - | 1 | 214 | 40029596 | ||
CR541921.1 | - | 1 | 169 | |||
JN591568.1 | - | 1 | 169 | |||
JN591569.1 | - | 1 | 169 | |||
JN591570.1 | - | 1 | 169 | |||
JN848522.1 | - | 1 | 169 | |||
M54886.1 | - | 1 | 229 | |||
V00544.1 | - | 1 | 229 | |||
V00548.1 | - | 1 | 151 | |||
V00549.1 | - | 1 | 229 |
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | E00175.1 | - | 1 | 153 |
Gene Symbol: IFNA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021268.2 | NP_067091.1 | 1 | 218 | ||
GenBank mRNA | BC096732.1 | - | 1 | 211 | 40015059 | |
BC098355.1 | - | 1 | 211 | 40015056 | ||
M11026.1 | - | 1 | 218 | |||
M38289.1 | - | 1 | 311 | |||
M71246.1 | - | 1 | 130 |
Gene Symbol: IFNA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002175.2 | NP_002166.2 | 1 | 217 | ||
GenBank mRNA | BC069329.1 | - | 1 | 177 | 7262150 | |
BC069372.1 | - | 1 | 177 | 7262162 | ||
BC069408.1 | - | 1 | 177 | 7262174 | ||
BC096699.1 | - | 1 | 211 | 40015057 | ||
BC101638.1 | - | 1 | 176 | 8069144 | ||
BC101640.1 | - | 1 | 176 | 8069146 | ||
M12350.1 | - | 1 | 217 | |||
M28586.1 | - | 1 | 217 | |||
V00540.1 | - | 1 | 177 | |||
X00145.1 | - | 1 | 217 |
Gene Symbol: IFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002169.2 | NP_002160.1 | 1 | 226 | ||
GenBank mRNA | BC093755.1 | - | 1 | 226 | 7939600 | |
BC093757.1 | - | 1 | 226 | 7939602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
