siRNA Details
Gene Symbol: IFNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000605.3 | NP_000596.2 | 1 | 339 | ||
GenBank mRNA | BC074936.2 | - | 1 | 315 | 30915269 | |
BC074937.2 | - | 1 | 315 | 30915492 | ||
BC104163.1 | - | 1 | 316 | 40029595 | ||
BC104164.1 | - | 1 | 316 | 40029596 | ||
CR541921.1 | - | 1 | 271 | |||
JN591569.1 | - | 1 | 271 | |||
JN848522.1 | - | 1 | 271 | |||
M54886.1 | - | 1 | 331 | |||
V00544.1 | - | 1 | 331 | |||
V00548.1 | - | 1 | 253 | |||
V00549.1 | - | 1 | 331 |
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | E00175.1 | - | 1 | 258 |
Gene Symbol: IFNA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002173.3 | NP_002164.1 | 1 | 280 | ||
GenBank mRNA | M28585.1 | - | 1 | 280 | ||
X00140.1 | - | 1 | 280 |
Gene Symbol: IFNA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002175.2 | NP_002166.2 | 1 | 322 | ||
GenBank mRNA | BC069329.1 | - | 1 | 282 | 7262150 | |
BC069372.1 | - | 1 | 282 | 7262162 | ||
BC069408.1 | - | 1 | 282 | 7262174 | ||
BC096699.1 | - | 1 | 316 | 40015057 | ||
BC101638.1 | - | 1 | 281 | 8069144 | ||
BC101640.1 | - | 1 | 281 | 8069146 | ||
M12350.1 | - | 1 | 322 | |||
M28586.1 | - | 1 | 322 | |||
V00540.1 | - | 1 | 282 | |||
X00145.1 | - | 1 | 322 |
Gene Symbol: IFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002169.2 | NP_002160.1 | 1 | 331 | ||
GenBank mRNA | BC093755.1 | - | 1 | 331 | 7939600 | |
BC093757.1 | - | 1 | 331 | 7939602 | ||
V00541.1 | - | 1 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
