siRNA Details
Gene Symbol: IFNA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002175.2 | NP_002166.2 | 1 | 500 | ||
GenBank mRNA | BC069329.1 | - | 1 | 460 | 7262150 | |
BC069372.1 | - | 1 | 460 | 7262162 | ||
BC069408.1 | - | 1 | 460 | 7262174 | ||
BC096699.1 | - | 1 | 494 | 40015057 | ||
BC101638.1 | - | 1 | 459 | 8069144 | ||
BC101640.1 | - | 1 | 459 | 8069146 | ||
M12350.1 | - | 1 | 500 | |||
M28586.1 | - | 1 | 500 | |||
V00540.1 | - | 1 | 460 | |||
X00145.1 | - | 1 | 500 |
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | E00175.1 | - | 1 | 436 |
Gene Symbol: IFNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002172.2 | NP_002163.2 | 1 | 496 | ||
GenBank mRNA | BC074956.2 | - | 1 | 496 | 30915281 | |
BC104159.1 | - | 1 | 494 | 40029547 | ||
BC104160.1 | - | 1 | 494 | 40029549 | ||
V00542.1 | - | 1 | 508 |
Gene Symbol: IFNA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002173.3 | NP_002164.1 | 1 | 458 | ||
GenBank mRNA | M28585.1 | - | 1 | 458 | ||
X00140.1 | - | 1 | 458 |
Gene Symbol: IFNA22P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036676.1 | 1 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
