Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000265.5 | NP_000256.4 | 2, 3 | 225 | ||
GenBank mRNA | AK223217.1 | - | 2, 3 | 176 | ||
AK292094.1 | - | 2, 3 | 176 | |||
AK316552.1 | - | 2, 3 | 176 | |||
BC002816.2 | - | 2, 3 | 225 | 3633829 | ||
BC065731.1 | - | 2, 3 | 181 | 6735930 |
Gene Symbol: NCF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298655.1 | - | 2, 3 | 176 |
Gene Symbol: NCF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003187.3 | 2, 3 | 208 | |||
GenBank mRNA | AF330627.1 | - | 2, 3 | 165 | ||
AK316329.1 | - | 2, 3 | 208 | |||
DC389713.1 | - | 2, 3 | 208 | |||
M25665.1 | - | 2, 3 | 167 | |||
M55067.1 | - | 2, 3 | 177 |