Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000265.5 | NP_000256.4 | 6 | 630 | ||
GenBank mRNA | AK127905.1 | - | 6 | 654 | ||
AK223217.1 | - | 6 | 581 | |||
AK292094.1 | - | 6 | 581 | |||
AK316341.1 | - | 4 | 339 | |||
AK316552.1 | - | 6 | 581 | |||
BC002816.2 | - | 6 | 630 | 3633829 | ||
BC065731.1 | - | 6 | 586 | 6735930 |