siRNA Details
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014.4 | NP_001005.1 | 5 | 541 | ||
NM_001203245.2 | NP_001190174.1 | 5 | 693 | |||
NM_001204091.1 | NP_001191020.1 | 5 | 545 | |||
GenBank mRNA | AB209165.1 | - | 1 | 496 | ||
AK311797.1 | - | 5 | 488 | |||
BC001032.2 | - | 5 | 466 | 3139393 | ||
BC001955.1 | - | 5 | 520 | 2905318 | ||
BC005012.1 | - | 5 | 520 | 3637658 | ||
BC070235.1 | - | 5 | 464 | 6453788 | ||
BC071946.1 | - | 5 | 476 | 6305105 | ||
BC073799.1 | - | 5 | 553 | 5517455 | ||
BE397113.1 | - | 5 | 491 | |||
BQ072109.1 | - | 1 | 661 | |||
BU956627.1 | - | 5 | 469 | |||
CD385233.1 | - | 5 | 543 | |||
U14972.1 | - | 1 | 463 |
Gene Symbol: RPS10P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025353.1 | - | 1 | 3075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
