Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198924.3 | NP_944606.2 | 4 | 909 | ||
GenBank mRNA | AF498998.1 | - | 4 | 909 | ||
AK126632.1 | - | 1 | 3434 | |||
BC073926.1 | - | 4 | 976 |