Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 11 | 1864 | ||
NM_001284290.1 | NP_001271219.1 | 9 | 1426 | |||
NM_001293104.1 | NP_001280033.1 | 10 | 1679 | |||
NM_001293105.1 | NP_001280034.1 | 9 | 1536 | |||
NR_120531.1 | 10 | 1779 | ||||
XM_005250297.3 | XP_005250354.1 | 10 | 1706 | |||
XM_011516113.1 | XP_011514415.1 | 10 | 1608 | |||
XM_011516114.1 | XP_011514416.1 | 10 | 1536 | |||
XM_017012091.1 | XP_016867580.1 | 9 | 1455 | |||
XM_017012092.1 | XP_016867581.1 | 9 | 1442 | |||
XM_017012093.1 | XP_016867582.1 | 9 | 1383 | |||
GenBank mRNA | AK096764.1 | - | 3 | 2122 | ||
AK223406.1 | - | 11 | 1763 | |||
AK300223.1 | - | 11 | 1653 | |||
AK303819.1 | - | 9 | 1323 | |||
BC014142.2 | - | 11 | 1734 | 4662011 | ||
BM719712.1 | - | 2 | 117 | |||
CD014092.1 | - | 10 | 1561 | |||
CD014093.1 | - | 9 | 1419 | |||
CD014094.1 | - | 10 | 1661 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027026.1 | 4 | 334 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 7 | 924 | |||
GenBank mRNA | BC034017.1 | - | 7 | 925 | ||
BC067351.1 | - | 5 | 622 |
Gene Symbol: LOC653080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Z70705.1 | - | 1 | 14 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 4 | 528 | |||
NR_029426.1 | 7 | 820 | ||||
GenBank mRNA | AK097091.1 | - | 8 | 1200 | ||
AK289832.1 | - | 7 | 902 | |||
AK315849.1 | - | 7 | 893 | |||
X75940.1 | - | 4 | 529 | |||
Z70701.1 | - | 1 | 14 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289851.1 | - | 7 | 822 |