siRNA Details
Gene Symbol: HIST2H2BE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003528.2 | NP_003519.1 | 1 | 156 | ||
GenBank mRNA | AK093747.1 | - | 1 | 156 | ||
AK289725.1 | - | 2 | 514 | |||
BC005827.2 | - | 1 | 126 | 2989788 | ||
BC069193.1 | - | 1 | 138 | 3936695 | ||
BC096121.2 | - | 1 | 114 | 40002353 | ||
BC098112.1 | - | 1 | 114 | 40014245 | ||
BC098289.1 | - | 1 | 114 | 40014249 | ||
BC107084.1 | - | 1 | 115 | 40014260 | ||
BC107085.1 | - | 1 | 115 | 40014264 | ||
CR541895.1 | - | 1 | 114 | |||
M60756.1 | - | 1 | 39 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
