siRNA Details
Gene Symbol: HIST2H2BA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027337.1 | 1 | 196 | |||
GenBank mRNA | BC106916.2 | - | 1 | 196 | ||
BC126904.1 | - | 1 | 197 |
Gene Symbol: HIST1H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003522.3 | NP_003513.1 | 1 | 187 | ||
GenBank mRNA | BC096120.1 | - | 1 | 187 | 40002352 | |
BC096123.1 | - | 1 | 187 | 40002357 |
Gene Symbol: HIST1H2BN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003520.3 | NP_003511.1 | 1 | 187 | ||
GenBank mRNA | AK312228.1 | - | 1 | 296 | ||
BC101411.1 | - | 1 | 187 | 40021691 | ||
BC101412.1 | - | 1 | 187 | 40021693 | ||
BC101413.1 | - | 1 | 187 | 40021695 |
Gene Symbol: HIST1H2BO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003527.4 | NP_003518.2 | 1 | 225 | ||
GenBank mRNA | BC106720.2 | - | 1 | 208 | 40014271 | |
BX647290.1 | - | 1 | 225 |
Gene Symbol: HIST2H2BC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036461.1 | 1 | 187 | |||
GenBank mRNA | AK299891.1 | - | 1 | 351 |
Gene Symbol: HIST2H2BE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003528.2 | NP_003519.1 | 1 | 229 | ||
GenBank mRNA | AK093747.1 | - | 1 | 229 | ||
AK289725.1 | - | 2 | 587 | |||
BC005827.2 | - | 1 | 199 | 2989788 | ||
BC069193.1 | - | 1 | 211 | 3936695 | ||
BC096121.2 | - | 1 | 187 | 40002353 | ||
BC098112.1 | - | 1 | 187 | 40014245 | ||
BC098289.1 | - | 1 | 187 | 40014249 | ||
BC107084.1 | - | 1 | 188 | 40014260 | ||
BC107085.1 | - | 1 | 188 | 40014264 | ||
CR541895.1 | - | 1 | 187 | |||
M60756.1 | - | 1 | 112 |
Gene Symbol: HIST2H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024599.4 | NP_001019770.1 | 1 | 237 | ||
NM_001161334.1 | NP_001154806.1 | 1 | 237 | |||
GenBank mRNA | AK289664.1 | - | 1 | 234 | ||
AK296916.1 | - | 1 | 237 | |||
AK299108.1 | - | 1 | 234 | |||
BC110793.1 | - | 1 | 220 | 5224812 | ||
DA042901.1 | - | 1 | 237 |
Gene Symbol: HIST3H2BB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175055.2 | NP_778225.1 | 1 | 210 | ||
GenBank mRNA | AK091220.1 | - | 1 | 210 | ||
BC100855.1 | - | 1 | 209 | 40002412 | ||
BC100856.1 | - | 1 | 209 | 40002414 | ||
BC100857.1 | - | 1 | 209 | 40002416 | ||
BC100858.1 | - | 1 | 209 | 40002418 | ||
BC137467.1 | - | 1 | 209 | 9021467 | ||
BC137468.1 | - | 1 | 209 | 9021468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
