Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002124.3 | NP_002115.2 | 1 | 116 | ||
GenBank mRNA | AK289463.1 | - | 1 | 85 | ||
AK291987.1 | - | 1 | 116 | |||
AY770514.1 | - | 1 | 22 | |||
AY961072.1 | - | 1 | 22 | |||
AY961073.1 | - | 1 | 22 | |||
BC033827.1 | - | 1 | 65 | 5216243 | ||
BC108922.1 | - | 1 | 77 | 40023604 | ||
DQ002917.1 | - | 1 | 22 | |||
DQ140279.1 | - | 1 | 82 | |||
HG974574.1 | - | 1 | 22 | |||
HG974596.1 | - | 1 | 22 | |||
HG974600.1 | - | 1 | 22 | |||
HM067861.1 | - | 1 | 22 | |||
HM067862.1 | - | 1 | 22 | |||
HM067863.1 | - | 1 | 22 | |||
KJ534866.1 | - | 1 | 22 | |||
L02545.1 | - | 1 | 36 | |||
M11161.1 | - | 1 | 22 | |||
M20430.1 | - | 1 | 65 | |||
M20504.1 | - | 1 | 57 | |||
M28583.1 | - | 1 | 61 | |||
M30179.1 | - | 1 | 22 | |||
M33600.1 | - | 1 | 92 | |||
X03069.1 | - | 1 | 84 |
Gene Symbol: HLA-DRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001023.2 | - | 1 | 82 | 3345335 | |
M16941.1 | - | 1 | 45 |
Gene Symbol: HLA-DRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021983.4 | NP_068818.4 | 1 | 93 | ||
GenBank mRNA | AF361548.1 | - | 1 | 31 | ||
AF361549.1 | - | 1 | 31 | |||
AK292151.1 | - | 1 | 73 | |||
BC005312.1 | - | 1 | 85 | 3933019 | ||
DQ284431.1 | - | 1 | 83 | |||
DQ284433.1 | - | 1 | 84 | |||
GQ891550.1 | - | 1 | 22 | |||
M16942.1 | - | 1 | 50 |
Gene Symbol: HLA-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292140.1 | - | 1 | 85 | ||
U66826.1 | - | 1 | 62 |
Gene Symbol: LOC105369230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HG974601.1 | - | 1 | 22 | ||
HM067848.1 | - | 1 | 22 |