Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205201.1 | NP_001192130.1 | 3 | 407 | ||
NM_001586.2 | NP_001577.1 | 3 | 401 | |||
XM_011531116.1 | XP_011529418.1 | 3 | 445 | |||
XM_011531117.1 | XP_011529419.1 | 4 | 514 | |||
XM_011531118.2 | XP_011529420.1 | 4 | 642 | |||
GenBank mRNA | BC026183.1 | - | 3 | 412 | 5266650 | |
DB457033.1 | - | 3 | 409 | |||
DQ655913.2 | - | 4 | 479 | |||
U93720.1 | - | 3 | 385 |