siRNA Details
Gene Symbol: HBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000559.2 | NP_000550.2 | 3 | 518 | ||
GenBank mRNA | BC010913.1 | - | 3 | 524 | 4133117 | |
BC020719.1 | - | 3 | 518 | 4732536 | ||
V00512.1 | - | 1 | 81 | |||
X55655.1 | - | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
