Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021268.2 | NP_067091.1 | 1 | 373 | ||
GenBank mRNA | BC096732.1 | - | 1 | 366 | 40015059 | |
BC098355.1 | - | 1 | 366 | 40015056 | ||
M11026.1 | - | 1 | 373 | |||
M38289.1 | - | 1 | 466 | |||
M71246.1 | - | 1 | 285 |
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | E00175.1 | - | 1 | 308 |
Gene Symbol: IFNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002171.2 | NP_002162.1 | 1 | 370 | ||
GenBank mRNA | BC069409.1 | - | 1 | 370 | 7262186 | |
BC103972.2 | - | 1 | 370 | 40015081 | ||
BC103973.1 | - | 1 | 370 | 40015082 | ||
V00551.1 | - | 1 | 370 |
Gene Symbol: IFNA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002175.2 | NP_002166.2 | 1 | 372 | ||
GenBank mRNA | BC069329.1 | - | 1 | 332 | 7262150 | |
BC069372.1 | - | 1 | 332 | 7262162 | ||
BC069408.1 | - | 1 | 332 | 7262174 | ||
BC096699.1 | - | 1 | 366 | 40015057 | ||
BC101638.1 | - | 1 | 331 | 8069144 | ||
BC101640.1 | - | 1 | 331 | 8069146 | ||
M12350.1 | - | 1 | 372 | |||
M28586.1 | - | 1 | 372 | |||
V00540.1 | - | 1 | 332 | |||
X00145.1 | - | 1 | 372 |
Gene Symbol: IFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021068.2 | NP_066546.1 | 1 | 392 | ||
GenBank mRNA | BC074965.2 | - | 1 | 344 | 30915507 | |
BC074966.2 | - | 1 | 344 | 30915288 | ||
BC113640.1 | - | 1 | 380 | 8322692 | ||
BC113642.1 | - | 1 | 380 | 8322694 | ||
M27318.1 | - | 1 | 464 |
Gene Symbol: IFNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021057.2 | NP_066401.2 | 1 | 364 | ||
GenBank mRNA | BC074991.2 | - | 1 | 362 | 30915521 | |
BC074992.2 | - | 1 | 362 | 30915307 | ||
BC113994.1 | - | 1 | 364 | 40084821 | ||
BC114000.1 | - | 1 | 364 | 40084820 | ||
M34913.1 | - | 1 | 364 |