Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP4S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128126.2 | NP_001121598.1 | 4 | 648 | ||
NM_001254726.1 | NP_001241655.1 | 4 | 648 | |||
NM_001254727.1 | NP_001241656.1 | 4 | 648 | |||
NM_001254728.1 | NP_001241657.1 | 4 | 1005 | |||
NM_001254729.1 | NP_001241658.1 | 4 | 523 | |||
NM_007077.4 | NP_009008.2 | 4 | 648 | |||
XM_005267293.4 | XP_005267350.1 | 5 | 1091 | |||
XM_011536371.2 | XP_011534673.1 | 5 | 661 | |||
XM_011536372.2 | XP_011534674.1 | 4 | 1055 | |||
GenBank mRNA | AB030654.1 | - | 4 | 468 | ||
AF155159.1 | - | 4 | 442 | |||
AK304115.1 | - | 4 | 469 | |||
AK316517.1 | - | 4 | 471 | |||
BC001259.1 | - | 4 | 431 | 3454117 | ||
BF671561.1 | - | 4 | 521 | |||
BT006701.1 | - | 3 | 259 | |||
BX247969.1 | - | 4 | 450 | |||
BX388185.2 | - | 4 | 434 | |||
CR457100.1 | - | 3 | 259 |