Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321525.1 | NP_001308454.1 | 19 | 2478 | ||
NM_001321526.1 | NP_001308455.1 | 19 | 2568 | |||
NM_001321527.1 | NP_001308456.1 | 19 | 2532 | |||
NM_001321528.1 | NP_001308457.1 | 19 | 2522 | |||
NM_001321529.1 | NP_001308458.1 | 18 | 2355 | |||
NM_001321530.1 | NP_001308459.1 | 18 | 2247 | |||
NM_207328.3 | NP_997211.2 | 19 | 2550 | |||
XM_017003424.1 | XP_016858913.1 | 19 | 2182 | |||
GenBank mRNA | AK122580.1 | - | 9 | 3974 | ||
AK131197.1 | - | 13 | 1587 | |||
AK298131.1 | - | 18 | 1927 | |||
AK302887.1 | - | 17 | 3282 | |||
BC068596.1 | - | 19 | 2221 | 4837765 |