Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000184.2 | NP_000175.1 | 2 | 355 | ||
GenBank mRNA | AF110493.1 | - | 2 | 249 | ||
AK290492.1 | - | 2 | 355 | |||
BC010914.1 | - | 2 | 359 | 4134713 | ||
BC029387.1 | - | 2 | 385 | 4762402 | ||
BC130457.1 | - | 2 | 350 | 40146488 | ||
BC130459.1 | - | 2 | 350 | 40146490 | ||
M15386.1 | - | 2 | 319 | |||
X55656.1 | - | 2 | 282 |
Gene Symbol: HBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005330.3 | NP_005321.1 | 2 | 555 | ||
GenBank mRNA | BC015537.2 | - | 4 | 647 | 3884290 | |
CR541912.1 | - | 2 | 302 |
Gene Symbol: HBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000559.2 | NP_000550.2 | 2 | 355 | ||
GenBank mRNA | BC010913.1 | - | 2 | 361 | 4133117 | |
BC020719.1 | - | 2 | 355 | 4732536 |