siRNA Details
Gene Symbol: DNAJB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005494.2 | NP_005485.1 | 3 | 268 | ||
NM_058246.3 | NP_490647.1 | 3 | 267 | |||
XM_005249515.2 | XP_005249572.1 | 3 | 259 | |||
XM_005249516.2 | XP_005249573.1 | 3 | 403 | |||
XM_006715823.1 | XP_006715886.1 | 3 | 259 | |||
XM_011515704.1 | XP_011514006.1 | 3 | 238 | |||
XM_017011633.1 | XP_016867122.1 | 3 | 280 | |||
GenBank mRNA | AB014888.1 | - | 3 | 213 | ||
AB015798.1 | - | 3 | 260 | |||
AB015799.1 | - | 3 | 260 | |||
AB209859.1 | - | 3 | 261 | |||
AF060703.1 | - | 3 | 258 | |||
AF075601.1 | - | 3 | 245 | |||
AK223601.1 | - | 3 | 278 | |||
AK291953.1 | - | 3 | 250 | |||
AK293025.1 | - | 3 | 251 | |||
AK297497.1 | - | 3 | 259 | |||
AK301152.1 | - | 3 | 280 | |||
AL136707.1 | - | 3 | 272 | |||
BC000177.2 | - | 3 | 259 | 2900421 | ||
BC002446.2 | - | 3 | 267 | 3346070 | ||
CR533498.1 | - | 2 | 113 | |||
EU075177.1 | - | 3 | 257 |
Gene Symbol: APOL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK297892.1 | - | 267 |
Gene Symbol: TMEM135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF080569.1 | - | 3 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
