siRNA Details
Gene Symbol: DNAJB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005494.2 | NP_005485.1 | 5 | 491 | ||
NM_058246.3 | NP_490647.1 | 5 | 490 | |||
XM_005249515.2 | XP_005249572.1 | 5 | 482 | |||
XM_005249516.2 | XP_005249573.1 | 5 | 626 | |||
XM_006715823.1 | XP_006715886.1 | 5 | 482 | |||
XM_011515704.1 | XP_011514006.1 | 5 | 461 | |||
XM_017011633.1 | XP_016867122.1 | 5 | 503 | |||
GenBank mRNA | AB014888.1 | - | 5 | 436 | ||
AB015798.1 | - | 5 | 483 | |||
AB015799.1 | - | 5 | 483 | |||
AB209859.1 | - | 5 | 484 | |||
AF060703.1 | - | 5 | 481 | |||
AF075601.1 | - | 5 | 468 | |||
AK223601.1 | - | 5 | 501 | |||
AK291953.1 | - | 5 | 473 | |||
AK293025.1 | - | 5 | 474 | |||
AK297796.1 | - | 4 | 381 | |||
AK297989.1 | - | 5 | 353 | |||
AL136707.1 | - | 5 | 495 | |||
BC000177.2 | - | 5 | 482 | 2900421 | ||
BC002446.2 | - | 5 | 490 | 3346070 | ||
CR533498.1 | - | 4 | 336 | |||
EU075177.1 | - | 5 | 480 |
Gene Symbol: APOL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK297892.1 | - | 490 |
Gene Symbol: TMEM135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF080569.1 | - | 5 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
